Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIR3
Entrez Gene ID
853605
Gene Name
Gene Aliases
HPC1, YJR140C
Chromosome Location
Chr.X: 690750 - 695696 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181798.1 NP_012674.1 1 - 1 4849 75

View Full Product Details