Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFH1
Entrez Gene ID
853771
Gene Name
Gene Aliases
YKL091C
Chromosome Location
Chr.XI: 269718 - 270650 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179657.1 NP_012832.1 1 - 1 855 78

View Full Product Details