Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OAC1
Entrez Gene ID
853739
Gene Name
Gene Aliases
YKL120W
Chromosome Location
Chr.XI: 217344 - 218318 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179686.1 NP_012802.1 1 - 1 754 96

View Full Product Details