Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLN1
Entrez Gene ID
853920
Gene Name
Gene Aliases
PET10, YKR046C
Chromosome Location
Chr.XI: 524223 - 525074 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179836.3 NP_012972.3 1 - 1 748 94

View Full Product Details