Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRC3
Entrez Gene ID
850624
Gene Name
Gene Aliases
YLL035W
Chromosome Location
Chr.XII: 68580 - 70478 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181855.1 NP_013065.1 1 - 1 1647 108

View Full Product Details