Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRL2
Entrez Gene ID
850771
Gene Name
Gene Aliases
YLR082C
Chromosome Location
Chr.XII: 292394 - 293572 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181969.1 NP_013183.1 1 - 1 1053 86

View Full Product Details