Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS31
Entrez Gene ID
850864
Gene Name
Gene Aliases
RPS37, UBI3, YLR167W
Chromosome Location
Chr.XII: 498947 - 499405 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182054.1 NP_013268.1 1 - 1 349 72

View Full Product Details