Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ENT2
Entrez Gene ID
850903
Gene Name
Gene Aliases
YLR206W
Chromosome Location
Chr.XII: 554578 - 556419 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182093.1 NP_013307.1 1 - 1 1442 87

View Full Product Details