Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDC25
Entrez Gene ID
851019
Gene Name
Gene Aliases
CTN1, YLR310C
Chromosome Location
Chr.XII: 752224 - 756993 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182198.1 NP_013413.1 1 - 1 4653 63

View Full Product Details