Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRX8
Entrez Gene ID
851079
Gene Name
Gene Aliases
YLR364W
Chromosome Location
Chr.XII: 854062 - 854391 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182253.3 NP_013468.3 1 - 1 248 93

View Full Product Details