Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UTP21
Entrez Gene ID
851125
Gene Name
Gene Aliases
YLR409C
Chromosome Location
Chr.XII: 934414 - 937233 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182297.3 NP_013513.3 1 - 1 2756 72

View Full Product Details