Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBX2
Entrez Gene ID
854995
Gene Name
Gene Aliases
SEL1, YML013W
Chromosome Location
Chr.XIII: 244149 - 245903 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182371.1 NP_013699.1 1 - 1 1648 83

View Full Product Details