Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSF2
Entrez Gene ID
854957
Gene Name
Gene Aliases
ECM6, YML048W
Chromosome Location
Chr.XIII: 178426 - 179637 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182405.1 NP_013664.1 1 - 1 850 77

View Full Product Details