Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SKY1
Entrez Gene ID
855256
Gene Name
Gene Aliases
YMR216C
Chromosome Location
Chr.XIII: 698811 - 701039 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182723.1 NP_013943.1 1 - 1 2150 79

View Full Product Details