Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SSN8
Entrez Gene ID
855706
Gene Name
Gene Aliases
CNC1, GIG3, NUT9, RYE2, SRB11, UME3, YNL025C
Chromosome Location
Chr.XIV: 584320 - 585291 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182864.3 NP_014373.3 1 - 1 849 102

View Full Product Details