Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHO2
Entrez Gene ID
855634
Gene Name
Gene Aliases
YNL090W
Chromosome Location
Chr.XIV: 456565 - 457143 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182928.3 NP_014309.3 1 - 1 452 75

View Full Product Details