Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYB5
Entrez Gene ID
855612
Gene Name
Gene Aliases
YNL111C
Chromosome Location
Chr.XIV: 416940 - 417302 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182949.3 NP_014288.3 1 - 1 250 101

View Full Product Details