Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GIM3
Entrez Gene ID
855569
Gene Name
Gene Aliases
PFD4, YNL153C
Chromosome Location
Chr.XIV: 345669 - 346058 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182991.1 NP_014246.1 1 - 1 249 85

View Full Product Details