Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YCK2
Entrez Gene ID
855568
Gene Name
Gene Aliases
YNL154C
Chromosome Location
Chr.XIV: 343636 - 345276 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182992.1 NP_014245.1 1 - 1 1548 94

View Full Product Details