Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC2
Entrez Gene ID
855449
Gene Name
Gene Aliases
YNL272C
Chromosome Location
Chr.XIV: 126805 - 129084 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183110.1 NP_014127.1 1 - 1 1847 87

View Full Product Details