Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AAD14
Entrez Gene ID
855385
Gene Name
Gene Aliases
YNL331C
Chromosome Location
Chr.XIV: 16118 - 17248 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183169.1 NP_014068.1 1 - 1 150 66

View Full Product Details