Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BUD17
Entrez Gene ID
855761
Gene Name
Gene Aliases
YNR027W
Chromosome Location
Chr.XIV: 674923 - 675876 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183204.1 NP_014424.1 1 - 1 750 67

View Full Product Details