Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
REX4
Entrez Gene ID
854075
Gene Name
Gene Aliases
YOL080C
Chromosome Location
Chr.XV: 180558 - 181427 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183334.1 NP_014561.1 1 - 1 650 64

View Full Product Details