Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPO21
Entrez Gene ID
854062
Gene Name
Gene Aliases
MPC70, YOL091W
Chromosome Location
Chr.XV: 145334 - 147163 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183345.1 NP_014550.1 1 - 1 649 84

View Full Product Details