Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADE2
Entrez Gene ID
854295
Gene Name
Gene Aliases
YOR128C
Chromosome Location
Chr.XV: 564476 - 566191 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183547.3 NP_014771.3 1 - 1 1557 75

View Full Product Details