Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DFR1
Entrez Gene ID
854411
Gene Name
Gene Aliases
YOR236W
Chromosome Location
Chr.XV: 780906 - 781541 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183655.1 NP_014879.1 1 - 1 547 91

View Full Product Details