Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MET7
Entrez Gene ID
854415
Gene Name
Gene Aliases
MET23, YOR241W
Chromosome Location
Chr.XV: 786995 - 788641 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183660.1 NP_014884.1 1 - 1 1547 100

View Full Product Details