Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EGD1
Entrez Gene ID
856070
Gene Name
Gene Aliases
YPL037C
Chromosome Location
Chr.XVI: 481428 - 481901 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183851.1 NP_015288.1 1 - 1 349 98

View Full Product Details