Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLR1
Entrez Gene ID
856014
Gene Name
Gene Aliases
LPG17, YPL091W
Chromosome Location
Chr.XVI: 375502 - 376953 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183905.1 NP_015234.1 1 - 1 1254 86

View Full Product Details