Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HOS3
Entrez Gene ID
855987
Gene Name
Gene Aliases
YPL116W
Chromosome Location
Chr.XVI: 329606 - 331699 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183930.1 NP_015209.1 1 - 1 1955 61

View Full Product Details