Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JID1
Entrez Gene ID
856174
Gene Name
Gene Aliases
YPR061C
Chromosome Location
Chr.XVI: 675977 - 676882 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184158.1 NP_015386.1 1 - 1 753 91

View Full Product Details