Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THI22
Entrez Gene ID
856239
Gene Name
Gene Aliases
YPR121W
Chromosome Location
Chr.XVI: 778583 - 780301 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184218.2 NP_015446.2 1 - 1 1555 77

View Full Product Details