Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SGE1
Entrez Gene ID
856327
Gene Name
Gene Aliases
NOR1, YPR198W
Chromosome Location
Chr.XVI: 934034 - 935665 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184295.1 NP_015524.1 1 - 1 1449 81

View Full Product Details