Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDT14
Entrez Gene ID
397008
Gene Name
nudix hydrolase 14
Gene Aliases
UGPP
Chromosome Location
Chr.Unknown: 3289576 - 3296920 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021081549.1 XP_020937208.1 - 302 67
XM_021081550.1 XP_020937209.1 - 305 67
XM_021081551.1 XP_020937210.1 - 295 67
XM_021081552.1 XP_020937211.1 - 488 67
XM_021081553.1 XP_020937212.1 - 331 67
XM_021081554.1 XP_020937213.1 - 338 67
XM_021081555.1 XP_020937214.1 - 302 67
XM_021081556.1 XP_020937215.1 - 303 67
GenBank mRNA AJ539380.1 - - 190 67

View Full Product Details