Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSF1
Entrez Gene ID
100513084
Gene Name
colony stimulating factor 1
Gene Aliases
MCSFBETA
Chromosome Location
Chr.4: 110178466 - 110198232 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244523.1 NP_001231452.1 4 - 5 559 145
XM_021088653.1 XP_020944312.1 - 904 145
XM_021088654.1 XP_020944313.1 - 660 145
GenBank mRNA AJ583506.1 - 2 - 3 231 145
DY405107.1 - 1 - 2 139 145

View Full Product Details