Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLR1C
Entrez Gene ID
414414
Gene Name
RNA polymerase I and III subunit C
Gene Aliases
RPA39
Chromosome Location
Chr.7: 38511521 - 38531938 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005666040.3 XP_005666097.1 - 1003 66
XM_005666041.3 XP_005666098.1 - 987 66
GenBank mRNA AK234320.1 - 8 - 9 948 66
AK238913.1 - 7 - 8 1093 66
AK391489.1 - 7 - 8 1100 66
AK399021.1 - 8 - 9 972 66
AY550050.1 - 1 - 2 30 66

View Full Product Details