Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMA4
Entrez Gene ID
397058
Gene Name
proteasome 20S subunit alpha 4
Gene Aliases
-
Chromosome Location
Chr.7: 47440576 - 47448599 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244468.1 NP_001231397.1 5 - 6 394 109
XM_021098292.1 XP_020953951.1 - 496 109
GenBank mRNA AK231641.1 - 5 - 6 402 109
AK389857.1 - 5 - 6 414 109
AK393796.1 - 5 - 6 415 109
AY610140.1 - 5 - 6 394 109
X91847.1 - - 179 109

View Full Product Details