Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WNT10B
Entrez Gene ID
100126276
Gene Name
Wnt family member 10B
Gene Aliases
-
Chromosome Location
Chr.5: 14997377 - 15013928 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287016.1 NP_001273945.1 4 - 4 1099 105
XM_021090858.1 XP_020946517.1 - 3890 105
XM_021090859.1 XP_020946518.1 - 3944 105
XM_021090860.1 XP_020946519.1 - 1969 105
XM_021090861.1 XP_020946520.1 - 1724 105
GenBank mRNA DQ518413.1 - 3 - 3 653 105
EU181371.1 - 4 - 4 917 105
KF569219.1 - 4 - 4 1099 105

View Full Product Details