Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDT2
Entrez Gene ID
397577
Gene Name
nudix hydrolase 2
Gene Aliases
-
Chromosome Location
Chr.10: 32450986 - 32466893 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214309.2 NP_999474.1 4 - 4 543 106
XM_005668056.3 XP_005668113.1 - 501 106
GenBank mRNA AK233880.1 - 3 - 3 470 106
AK391821.1 - 3 - 3 458 106
U38619.1 - 4 - 4 546 106

View Full Product Details