Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTH
Entrez Gene ID
399502
Gene Name
parathyroid hormone
Gene Aliases
-
Chromosome Location
Chr.2: 45757398 - 45760433 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214401.1 NP_999566.1 3 - 3 353 89
XM_021081593.1 XP_020937252.1 - 653 89
GenBank mRNA X05722.1 - - 353 89

View Full Product Details