Assay Design
The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP4A24
- Entrez Gene ID
-
403326
- Gene Name
- cytochrome P450 4A24
- Gene Aliases
-
CYP4A25, CYPIVA24
- Chromosome Location
-
Chr.6: 164496928 - 164518189 on Build Sscrofa11.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_214424.1 |
NP_999589.1 |
8 - 8 |
949 |
87 |
- Gene Symbol
- LOC100737897
- Entrez Gene ID
-
100737897
- Gene Name
- cytochrome P450 4A24
- Gene Aliases
-
CYP4A24, Cyp4A
- Chromosome Location
-
Chr.6: 164620278 - 164641079 on Build Sscrofa11.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF384031.1 |
- |
8 - 8 |
949 |
87 |
- Gene Symbol
- LOC110255311
- Entrez Gene ID
-
110255311
- Gene Name
- cytochrome P450 4A24-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 164523289 - 164544414 on Build Sscrofa11.1
- Gene Symbol
- LOC110261409
- Entrez Gene ID
-
110261409
- Gene Name
- cytochrome P450 4A25-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 164340816 - 164346993 on Build Sscrofa11.1
View Full Product Details