Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PNPLA2
Entrez Gene ID
100049704
Gene Name
patatin like phospholipase domain containing 2
Gene Aliases
ATGL
Chromosome Location
Chr.2: 513065 - 517962 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098605.1 NP_001092075.1 8 - 8 1146 62
XM_021076533.1 XP_020932192.1 - 1384 62
GenBank mRNA EF567018.1 - 8 - 8 1146 62
EF583921.1 - 8 - 8 1179 62
EF648448.1 - 8 - 8 1290 62
EU009401.2 - 8 - 8 1290 62
EU047807.1 - 8 - 8 1149 62
EU373817.1 - 8 - 8 1146 62

View Full Product Details