Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAG1
Entrez Gene ID
397506
Gene Name
recombination activating 1
Gene Aliases
RAG-1
Chromosome Location
Chr.2: 24565447 - 24573853 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123184.1 NP_001116656.1 1 - 1 3045 87

View Full Product Details