Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FSCN3
Entrez Gene ID
733598
Gene Name
fascin actin-bundling protein 3
Gene Aliases
-
Chromosome Location
Chr.18: 20684621 - 20693552 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044547.1 NP_001038012.1 4 - 5 1222 73
GenBank mRNA DQ324422.1 - 4 - 5 1222 73

View Full Product Details