Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LY86
Entrez Gene ID
396639
Gene Name
lymphocyte antigen 86
Gene Aliases
MD-1
Chromosome Location
Chr.7: 4108124 - 4156629 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097415.1 NP_001090884.1 1 - 2 179 71
GenBank mRNA AB190766.1 - 1 - 2 179 71
AK231170.1 - 1 - 2 171 71
AK237721.1 - 1 - 2 165 71
AK346556.1 - 1 - 2 151 71
AK346608.1 - 1 - 2 172 71
AK346874.1 - 1 - 2 158 71
AK346937.1 - 1 - 2 168 71

View Full Product Details