Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NEU1
Entrez Gene ID
100124381
Gene Name
neuraminidase 1
Gene Aliases
-
Chromosome Location
Chr.7: 23953721 - 23958659 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101822.1 NP_001095292.1 4 - 5 936 81

View Full Product Details