Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TOR1A
Entrez Gene ID
100125953
Gene Name
torsin family 1 member A
Gene Aliases
DYT1
Chromosome Location
Chr.1: 269975919 - 269985478 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105298.1 NP_001098768.1 2 - 3 452 133
GenBank mRNA AK233486.1 - 2 - 3 513 133
EF101283.1 - 2 - 3 452 133
EF101284.1 - 2 - 3 452 133

View Full Product Details