Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STC2
Entrez Gene ID
100126236
Gene Name
stanniocalcin 2
Gene Aliases
-
Chromosome Location
Chr.16: 50989676 - 51001412 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110173.1 NP_001103643.1 1 - 2 173 57
GenBank mRNA AK349078.1 - 1 - 2 487 57
EU165323.1 - 1 - 2 173 57

View Full Product Details