Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POU5F1
Entrez Gene ID
100127461
Gene Name
POU class 5 homeobox 1
Gene Aliases
OCT3, OCT4, Oct-4
Chromosome Location
Chr.7: 23564654 - 23570691 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113060.1 NP_001106531.1 3 - 4 663 86
XM_021097869.1 XP_020953528.1 - 757 86
GenBank mRNA KJ023671.1 - 1 - 2 1001 86
KJ023672.1 - 2 - 3 690 86

View Full Product Details