Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C17H20orf85
Entrez Gene ID
100144513
Gene Name
chromosome 17 C20orf85 homolog
Gene Aliases
C20orf85
Chromosome Location
Chr.17: 58373168 - 58383135 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123088.1 NP_001116560.1 3 - 4 311 68

View Full Product Details